Learn More
Medchemexpress LLC HY-107658 5mg Medchemexpress, SN 6 CAS:415697-08-4 Purity:>98%
SDP
Catalog No. 501968450
Encompass_Preferred
Medchemexpress, HY-107658 5mg SN 6 CAS:415697-08-4 SN 6 is a selective Na+/Ca2+ exchanger (NCX) inhibitor, and inhibits 45Ca2+ uptake by NCX1, NCX2, and NCX3, with IC50s of 2.9, 16, and 8.6 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8450 Supplier Medchemexpress LLC Supplier No. HY1076585MG
Medchemexpress, HY-107658 5mg SN 6 CAS:415697-08-4 SN 6 is a selective Na+/Ca2+ exchanger (NCX) inhibitor, and inhibits 45Ca2+ uptake by NCX1, NCX2, and NCX3, with IC50s of 2.9, 16, and 8.6 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.