Learn More
Cayman Chemical sn-glycero-3-Phosphocholine, Purity: ≥95%, CAS: 28319-77-9, 25 g
SDP
Catalog No. 502984928
Encompass_Preferred
A nootropic phospholipid; a precursor to choline biosynthesis and an intermediate in the metabolism of phosphatidylcholine; increases acetylcholine synthesis and release in rat hippocampus; prevents scopolamine-induced amnesia in a passive avoidance test in rats; exerts neuroprotective effects in models of altered cholinergic neurotransmission and models of brain vascular injury
Catalog No. 50-298-4928 Supplier Cayman Chemical Supplier No. 2073625 G
A nootropic phospholipid; a precursor to choline biosynthesis and an intermediate in the metabolism of phosphatidylcholine; increases acetylcholine synthesis and release in rat hippocampus; prevents scopolamine-induced amnesia in a passive avoidance test in rats; exerts neuroprotective effects in models of altered cholinergic neurotransmission and models of brain vascular injury