Learn More
Apexbio Technology LLC sn-Glycero-3-phosphocholine 28319-77-9 10g
SDP
Catalog No. 5000089604
Encompass_Preferred
sn-Glycero-3-phosphocholine (CAS 28319-77-9) also known as alpha-GPC or choline alfoscerate is a naturally occurring choline compound found in the brain and mammalian milk It serves as a precursor to acetylcholine a key neurotransmitter involved in cognitive processes by efficiently crossing the blood-brain barrier and supplying choline for neurotransmitter synthesis Preclinical and clinical research has examined its potential to enhance cognitive function particularly in neurodegenerative conditions such as Alzheimer s disease and vascular dementia Clinical studies have reported improved cognitive recovery in post-stroke and dementia patients following alpha-GPC administration supporting its utility in neuropharmacology and cognitive disorder research
Catalog No. 50-000-89604 Supplier Apexbio Technology LLC Supplier No. B115710000
sn-Glycero-3-phosphocholine (CAS 28319-77-9) also known as alpha-GPC or choline alfoscerate is a naturally occurring choline compound found in the brain and mammalian milk It serves as a precursor to acetylcholine a key neurotransmitter involved in cognitive processes by efficiently crossing the blood-brain barrier and supplying choline for neurotransmitter synthesis Preclinical and clinical research has examined its potential to enhance cognitive function particularly in neurodegenerative conditions such as Alzheimer s disease and vascular dementia Clinical studies have reported improved cognitive recovery in post-stroke and dementia patients following alpha-GPC administration supporting its utility in neuropharmacology and cognitive disorder research