Learn More
Medchemexpress LLC Sn-Glycero-3-phosphocholine | 28319-77-9 | 98.0% | 257.22 | 500 MG
SDP
Catalog No. 5000386275
Encompass_Preferred
sn-Glycero-3-phosphocholine (Choline Alfoscerate) is a crucial precursor in the biosynthesis of brain phospholipids, enhancing choline bioavailability in nervous tissue. This compound has demonstrated significant effects on cognitive function, offering a good safety profile and tolerability, and is effective in the treatment of Alzheimer's disease and dementia.
  • Precursor in brain phospholipid biosynthesis
  • Increases choline bioavailability
  • Supports cognitive function
  • Effective in treating Alzheimer's disease and dementia
  • Facilitates learning and memory
  • Improves brain transduction mechanisms
Catalog No. 50-003-86275 Supplier Medchemexpress LLC Supplier No. HY17552500MG
sn-Glycero-3-phosphocholine (Choline Alfoscerate) is a crucial precursor in the biosynthesis of brain phospholipids, enhancing choline bioavailability in nervous tissue. This compound has demonstrated significant effects on cognitive function, offering a good safety profile and tolerability, and is effective in the treatment of Alzheimer's disease and dementia.
  • Precursor in brain phospholipid biosynthesis
  • Increases choline bioavailability
  • Supports cognitive function
  • Effective in treating Alzheimer's disease and dementia
  • Facilitates learning and memory
  • Improves brain transduction mechanisms