Learn More
Medchemexpress LLC Sn-glycero-3-phosphocholine-d9 | 2260669-07-4 | 99.6% | 266.28 | 1 MG
SDP
Catalog No. 5000386375
Encompass_Preferred
sn-Glycero-3-phosphocholine-d9 is a deuterium-labeled form of sn-Glycero-3-phosphocholine, also known as Choline Alfoscerate. This compound acts as a precursor in the biosynthesis of brain phospholipids, enhancing choline bioavailability in nervous tissue. It has shown significant effects on cognitive function, with a good safety profile, making it effective in treating Alzheimer's disease and dementia.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-86375 Supplier Medchemexpress LLC Supplier No. HY17552S1MG
sn-Glycero-3-phosphocholine-d9 is a deuterium-labeled form of sn-Glycero-3-phosphocholine, also known as Choline Alfoscerate. This compound acts as a precursor in the biosynthesis of brain phospholipids, enhancing choline bioavailability in nervous tissue. It has shown significant effects on cognitive function, with a good safety profile, making it effective in treating Alzheimer's disease and dementia.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.