Learn More
Medchemexpress LLC sn-Glycero-3-phosphocholine (Standard) | 28319-77-9 | 99.7% | 257.22 | 100 MG
SDP
Catalog No. 5000386273
Encompass_Preferred
sn-Glycero-3-phosphocholine (Standard) is an analytical standard intended for research and analytical applications. It functions as a precursor in the biosynthesis of brain phospholipids, increasing the bioavailability of choline in nervous tissue. This compound demonstrates significant effects on cognitive function with a good safety profile and tolerability, and is effective in the treatment of Alzheimer's disease and dementia.
  • Intended for research and analytical applications.
  • Acts as a precursor in the biosynthesis of brain phospholipids.
  • Increases the bioavailability of choline in nervous tissue.
  • Demonstrates significant effects on cognitive function with good safety and tolerability.
  • Effective in the treatment of Alzheimer's disease and dementia.
  • Commonly used in qualitative, quantitative, and methodological research experiments.
  • Suitable for use in HPLC, GC, and MS applications.
Catalog No. 50-003-86273 Supplier Medchemexpress LLC Supplier No. HY17552R100MG
sn-Glycero-3-phosphocholine (Standard) is an analytical standard intended for research and analytical applications. It functions as a precursor in the biosynthesis of brain phospholipids, increasing the bioavailability of choline in nervous tissue. This compound demonstrates significant effects on cognitive function with a good safety profile and tolerability, and is effective in the treatment of Alzheimer's disease and dementia.
  • Intended for research and analytical applications.
  • Acts as a precursor in the biosynthesis of brain phospholipids.
  • Increases the bioavailability of choline in nervous tissue.
  • Demonstrates significant effects on cognitive function with good safety and tolerability.
  • Effective in the treatment of Alzheimer's disease and dementia.
  • Commonly used in qualitative, quantitative, and methodological research experiments.
  • Suitable for use in HPLC, GC, and MS applications.