Learn More
Medchemexpress LLC Sn-Glycerol 3-phosphate-13C3 disodium | 17989-41-2 | 98.7% | 219.02 | 1 MG
SDP
Catalog No. 5000437327
Encompass_Preferred
sn-Glycerol 3-phosphate-13C3 disodium is the 13C-labeled sn-Glycerol 3-phosphate disodium. sn-Glycerol 3-phosphate disodium is produced by the cytosolic glycerol 3-phosphate dehydrogenase pathway through the reduction of dihydroxyacetone phosphate using NADH formed during glycolysis.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-37327 Supplier Medchemexpress LLC Supplier No. HY113128S1MG
sn-Glycerol 3-phosphate-13C3 disodium is the 13C-labeled sn-Glycerol 3-phosphate disodium. sn-Glycerol 3-phosphate disodium is produced by the cytosolic glycerol 3-phosphate dehydrogenase pathway through the reduction of dihydroxyacetone phosphate using NADH formed during glycolysis.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.