Learn More
Medchemexpress LLC sn-Glycerol 3-phosphate | 17989-41-2 | 98.0% | 172.07 | 1 MG
SDP
Catalog No. 5000435753
Encompass_Preferred
sn-Glycerol 3-phosphate is a crucial intermediate in glycolysis and lipid metabolism. It is generated from dihydroxyacetone phosphate (DHAP) by the cytosolic sn-Glycerol 3-phosphate dehydrogenase (GPDH). Glyceroneogenesis is a significant supplier of sn-Glycerol 3-phosphate for lipid metabolism.
  • Intermediate in glycolysis and lipid metabolism.
  • Generated from dihydroxyacetone phosphate by cytosolic GPDH.
  • Passes through the permeable mitochondrial outer membrane.
  • Reoxidized to DHAP by mitochondrial FAD-GPDH.
  • DHAP subsequently channeled back to the cytosolic compartment.
Catalog No. 50-004-35753 Supplier Medchemexpress LLC Supplier No. HY1131281MG
sn-Glycerol 3-phosphate is a crucial intermediate in glycolysis and lipid metabolism. It is generated from dihydroxyacetone phosphate (DHAP) by the cytosolic sn-Glycerol 3-phosphate dehydrogenase (GPDH). Glyceroneogenesis is a significant supplier of sn-Glycerol 3-phosphate for lipid metabolism.
  • Intermediate in glycolysis and lipid metabolism.
  • Generated from dihydroxyacetone phosphate by cytosolic GPDH.
  • Passes through the permeable mitochondrial outer membrane.
  • Reoxidized to DHAP by mitochondrial FAD-GPDH.
  • DHAP subsequently channeled back to the cytosolic compartment.