Learn More
Medchemexpress LLC Sn-Glycerol 3-phosphate | 17989-41-2 | >=98.0% | 172.07 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11312810MM/1ML
Encompass_Preferred
sn-Glycerol 3-phosphate is a crucial intermediate involved in glycolysis and lipid metabolism. It is produced by the cytosolic sn-Glycerol 3-phosphate dehydrogenase (GPDH) from dihydroxyacetone phosphate (DHAP) and is also a key supplier for lipid metabolism through glyceroneogenesis.
  • Important intermediate in glycolysis and lipid metabolism.
  • Generated from dihydroxyacetone phosphate (DHAP) by cytosolic sn-Glycerol 3-phosphate dehydrogenase (GPDH).
  • Plays a role in glyceroneogenesis as a supplier for lipid metabolism.
  • Stable salt forms, such as sn-Glycerol 3-phosphate lithium, retain similar biological activity.
Catalog No. 50-004-36141