Learn More
TARGETMOL CHEMICALS INC sn-Glycerol 3-phosphate 1MG
SDP
Catalog No. 502876865
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.sn-Glycerol 3-phosphate, produced by the reduction of dihydroxyacetone phosphate by NADH formed during glycolysis of the cytoplasmic glycerol 3-phosphate dehydrogenase pathway. Purity 98.41%
Catalog No. 50-287-6865 Supplier TARGETMOL CHEMICALS INC Supplier No. T193431MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.sn-Glycerol 3-phosphate, produced by the reduction of dihydroxyacetone phosphate by NADH formed during glycolysis of the cytoplasmic glycerol 3-phosphate dehydrogenase pathway. Purity 98.41%