Learn More
Medchemexpress LLC HY-113128A 5mg Medchemexpress, sn-Glycerol 3-phosphate (lithium) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY113128A5MG
Encompass_Preferred
Medchemexpress, HY-113128A 5mg sn-Glycerol 3-phosphate (lithium) CAS: sn-Glycerol 3-phosphate lithium is produced by cytosolic glycerol 3-phosphate dehydrogenase pathway through the reduction of dihydroxyacetone phosphate using NADH formed during glycolysis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0836