Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SN38-COOH | 2170118-23-5 | 96.1% | 492.48 | C26H24N2O8
SDP

Catalog No. 5000203628
Encompass_Preferred

SN38-COOH is a carboxylated derivative of SN-38 used as a payload/linker building block for antibody-drug conjugate (ADC) synthesis. SN-38 is the active metabolite of the topoisomerase I inhibitor irinotecan and inhibits DNA and RNA synthesis.

  • Purity: 96.1%.
  • Molecular weight: 492.48 g/mol.
  • Formula: C26H24N2O8.
  • Appearance: solid.
  • Recommended storage: powder at -20°C (stable for 3 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Available pack sizes: 25 mg, 50 mg, 100 mg, 250 mg, 500 mg, 1 g, 5 g, 10 g.

Catalog No. 50-002-03628 Supplier Medchemexpress LLC Supplier No. HY1530135MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

SN38-COOH is a carboxylated derivative of SN-38 used as a payload/linker building block for antibody-drug conjugate (ADC) synthesis. SN-38 is the active metabolite of the topoisomerase I inhibitor irinotecan and inhibits DNA and RNA synthesis.

  • Purity: 96.1%.
  • Molecular weight: 492.48 g/mol.
  • Formula: C26H24N2O8.
  • Appearance: solid.
  • Recommended storage: powder at -20°C (stable for 3 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Available pack sizes: 25 mg, 50 mg, 100 mg, 250 mg, 500 mg, 1 g, 5 g, 10 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.