Learn More
Medchemexpress LLC Sn40 hydrochloride | 2768663-15-4 | 98.0% | 332.82 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY146241B1MG
Encompass_Preferred
SN40 hydrochloride is a potent amino acid transport (AAT) inhibitor, suitable for researching anticancer applications. It exhibits various Ki values against rat and human ASCT2, and EAAT1, EAAT2, EAAC1, and EAAT5.
  • Potent amino acid transport (AAT) inhibitor
  • Suitable for anticancer research
  • Targets multiple ASCT2 and EAAT isoforms
Catalog No. 50-003-60749