Learn More
Medchemexpress LLC HY-P0151 1mg Medchemexpress, SN50 CAS:213546-53-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP01511MG
Encompass_Preferred
Medchemexpress, HY-P0151 1mg SN50 CAS:213546-53-3 SN50 is a cell permeable inhibitor of NF-κB translocation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3023