Learn More
Medchemexpress LLC HY-P3229 1mg , SN52 CAS:1071173-56-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP32291MG
Encompass_Preferred
HY-P3229 1mg SN52 CAS: 1071173-56-2 SN52 is a potent, competitive, and cell-permeable inhibitor of NF-κB2. SN52 is a variant of the SN50 peptide and inhibits the nuclear translocation of p52-RelB heterodimers. SN52 has a strong radiosensitization effect on prostate cancer cells. SN52 can be used for cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1214