Learn More
Vector Laboratories Sambucus Nigra Lectin (SNA, EBL), CY3 CL-1303-1, Size: 1mg
SDP
Supplier:  Vector Laboratories CL13031
Encompass
Unit Size: 1 mg,Applications: Immunofluorescence, Glycobiology, Recommended Usage: The recommended concentration range for use is 5-20 µg/ml. Recommended Storage: 2-8 °C, Maximum Excitation: 552 nm, Maximum Emission: 565 nm, Solution: 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM CaCl2, and a proprietary stabilizer. Concentration: 1 mg active conjugate/ml, Conjugate: Cy3, Color of Fluorescence: Red, Sugar Specificity: Sialic Acid
Catalog No. 50-204-5634