Learn More
New England Biolabs, Inc. SnaBI – 2500 units
SDP
Catalog No. 50811978
Encompass
A restriction endonuclease that recognizes the sequence TAC_^GTA.
Catalog No. 50-811-978 Supplier New England Biolabs, Inc. Supplier No. R0130L
A restriction endonuclease that recognizes the sequence TAC_^GTA.