Learn More
Aviva Systems Biology Corp SNAG1 Antibody - N-terminal region (ARP52437_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52437_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNAG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SNAG1. Alternate names: FLJ11997, FLJ32560, MGC150827, MGC150829, SH3PX2, SH3PXD3B, SNX18, SNAG1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast
Catalog No. 50-014-60742