Learn More
Aviva Systems Biology Corp SNAG1 Antibody - middle region (ARP58951_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58951_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNAG1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SNAG1. Alternate names: FLJ11997, FLJ32560, FLJ61062, MGC150827, MGC150829, SH3PX2, SH3PXD3B, SNAG1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-538