Learn More
Aviva Systems Biology Corp SNAI1 Antibody - N-terminal region (ARP33314_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33314_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNAI1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SNAI1. Alternate names: SNA, SNAH, SNAIL, SLUGH2, SNAIL1, dJ710H13.1ApplicationIHC, WB. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-54958