Learn More
Aviva Systems Biology Corp SNAI2 Antibody - middle region (ARP33302_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33302_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNAI2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SNAI2. Alternate names: MGC10182, SLUG, SLUGH1, WS2D, SNAIL2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-014-54951