Learn More
Aviva Systems Biology Corp Snai3 Antibody - N-terminal region (ARP37207_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37207_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Snai3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the n terminal region of mouse Snai3. Alternate names: AI643946, Smuc, Zfp293ApplicationWB. Species reactivity: Cow, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-593