Learn More
Innovative Research Inc Snake Venom Phosphodiesterase I Purified lyophilized 100un
Catalog No. 502036470
Encompass_Preferred
Snake Venom Phosphodiesterase I Purified lyophilized from Innovative Research is prepared by process of Williams, Sung and Laskowski, JBC, 236, 1130 (1961) and treated to inactivate contaminating 5’-nucleotidase activity according to Sulkowski and Laskowski, Biochim. Biophys. Acta, 240, 443 (1961). This is a lyophilized in vials with a concentration of 20 Units/mg dry weight.
This product is useful successively hydrolyzing 5’-mononucleotides from 3’-OH-terminated riboand deoxyribo-oligonucleotides. The enzyme has an optimal pH range of 9.8-10.4 and a molecular weight of 115 kDa. Phosphodiesterase is inhibited by reducing agents such as glutathione, The enzyme has an optimal pH range of 9.8-10.4 and a molecular weight of 115 kDa. It is inhibited by reducing agents such as glutathione, cysteine and ascorbic acids and completely inhibited by 5 mM EDTA. ATP, ADP and AMP are partial inhibitors.
Catalog No. 50-203-6470 Supplier Innovative Research Inc Supplier No. IDBRPDEILY100UN
Add to Cart
Add to Cart
Snake Venom Phosphodiesterase I Purified lyophilized from Innovative Research is prepared by process of Williams, Sung and Laskowski, JBC, 236, 1130 (1961) and treated to inactivate contaminating 5’-nucleotidase activity according to Sulkowski and Laskowski, Biochim. Biophys. Acta, 240, 443 (1961). This is a lyophilized in vials with a concentration of 20 Units/mg dry weight.
This product is useful successively hydrolyzing 5’-mononucleotides from 3’-OH-terminated riboand deoxyribo-oligonucleotides. The enzyme has an optimal pH range of 9.8-10.4 and a molecular weight of 115 kDa. Phosphodiesterase is inhibited by reducing agents such as glutathione, The enzyme has an optimal pH range of 9.8-10.4 and a molecular weight of 115 kDa. It is inhibited by reducing agents such as glutathione, cysteine and ascorbic acids and completely inhibited by 5 mM EDTA. ATP, ADP and AMP are partial inhibitors.