Learn More
Enzo Life Sciences SNAP (20mg). CAS: 79032-48-7
SDP
Catalog No. 502010451
Encompass_Preferred
Stable analog of endogenous S-nitroso compounds. Nitric oxide (NO) donor and a source of NO in vivo which spontaneously releases NO under physiological conditions. The half-life time is 4.6 hours in aqueous media. Potent vasodilator in vitro and in vivo that does not induce pharmacological tolerance. Relaxes isolated bovine coronary artery rings with an EC50=0.1µM. It also inhibits vascular smooth muscle cell mitogenesis and proliferation, inhibits leukocyte adherence to endothelium, and inhibits cysteine proteases. The unique NO releasing properties of SNAP make it an ideal agent for studying the pharmacological and physiological actions of NO. Alternative name: S-Nitroso-N-acetyl-D,L-penicillamine. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) 100% ethanol (25mg/ml) or methanol; slightly soluble in water (2mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0451 Supplier Enzo Life Sciences Supplier No. BMLCN2100020
Stable analog of endogenous S-nitroso compounds. Nitric oxide (NO) donor and a source of NO in vivo which spontaneously releases NO under physiological conditions. The half-life time is 4.6 hours in aqueous media. Potent vasodilator in vitro and in vivo that does not induce pharmacological tolerance. Relaxes isolated bovine coronary artery rings with an EC50=0.1µM. It also inhibits vascular smooth muscle cell mitogenesis and proliferation, inhibits leukocyte adherence to endothelium, and inhibits cysteine proteases. The unique NO releasing properties of SNAP make it an ideal agent for studying the pharmacological and physiological actions of NO. Alternative name: S-Nitroso-N-acetyl-D,L-penicillamine. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) 100% ethanol (25mg/ml) or methanol; slightly soluble in water (2mg/ml). Long Term Storage: -20°C.