Learn More
Enzo Life Sciences SNAP-25 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210279R100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 195-206 (A195NQRATKML GSG206) of rat SNAP-25 (synaptosomal-associated protein 25). UniProt ID: P60881. Species reactivity: Mouse, Rat Zebrafish, Bovine. Applications: ICC, WB. Recommended Dilutions: Immunocytochemistry (1:100) Western Blot (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope-affinity purified IgG. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8102