Learn More
Anton Paar Snap 51 Advanced Portable Alcohol Meter for sugar-free distilled spirits
Supplier:  Anton Paar 183055
Encompass_Preferred
The Snap 51 portable alcohol meter measures the Proof concentration for quick and on-site alcohol checks in the industrial production of spirits. Only 2 mL of sample are required for a measurement, performed based on the oscillating U-tube technology. An RFID interface for quick sample identification and a Bluetooth interface for convenient data handling are integral parts of the instrument.Alcohol measuring range from 0 %v/v to 100 %v/vAccuracy of 0.1% v/vAmbient temperature suitable for -10°C up to +50°CTemperature correction of results is done automatically: no need for temperature correction tablesValues are shown in %v/v (corrected to 20 °C) or °Proof (corrected to 60 °F) or °Brix or °PlatoCalibration and adjustments are easy to perform with deionized water, following the predefined routine2 mL of sample are taken directly from the storage container with the help of the built-in pump.Includes Carrying Case.Recommended Accessories: NC1818217, NC1818220
Catalog No. NC1818209
Add to Cart
Request a Quote