Learn More
Medchemexpress LLC HY-107625A 25mg Medchemexpress, SNAP 94847 (hydrochloride) CAS:1781934-47-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107625A25MG
Encompass_Preferred
Medchemexpress, HY-107625A 25mg SNAP 94847 (HCl) CAS:1781934-47-1 SNAP 94847 hydrochloride is a novel, high affinity selective melanin-concentrating hormonereceptor1 (MCHR1) antagonist with (Ki= 2.2 nM, Kd=530 pM), it displays >80-fold and >500-fold selectivity over MCHα1A and MCHD2 receptors respectively. SNAP 94847 hydrochloride binds with high affinity to the mouse and rat MCHR1 with minimal cross-reactivity to other GPCR, ion channels, enzymes, and transporters[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8443