Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Snap 94847 hydrochloride | 1781934-47-1 | >98.0% | 515.03 g/mol | C29H32F2N2O2·HCl | 1 ML
SDP

Catalog No. 5000241622
Encompass_Preferred

SNAP 94847 hydrochloride is a potent, selective melanin-concentrating hormone receptor 1 (MCH1) antagonist supplied as a concentrated research solution. It is used to probe MCH1 signaling in neuroscience studies and has demonstrated effects on appetite-related behavior, anxiety, and hippocampal neurogenesis in preclinical models.

  • Potent MCH1 antagonist with Ki = 2.2 nM and Kd = 530 pM.
  • High selectivity over α1A and D2 receptors (>80-fold and >500-fold).
  • Provided as a 10 mM solution in DMSO for laboratory use.
  • Purity ≥98.0% by HPLC.
  • Molecular weight 515.03 g/mol; formula C29H32F2N2O2·HCl.
  • Store desiccated at room temperature; handle solvents per safety guidance.

Catalog No. 50-002-41622 Supplier Medchemexpress LLC Supplier No. HY107625A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SNAP 94847 hydrochloride is a potent, selective melanin-concentrating hormone receptor 1 (MCH1) antagonist supplied as a concentrated research solution. It is used to probe MCH1 signaling in neuroscience studies and has demonstrated effects on appetite-related behavior, anxiety, and hippocampal neurogenesis in preclinical models.

  • Potent MCH1 antagonist with Ki = 2.2 nM and Kd = 530 pM.
  • High selectivity over α1A and D2 receptors (>80-fold and >500-fold).
  • Provided as a 10 mM solution in DMSO for laboratory use.
  • Purity ≥98.0% by HPLC.
  • Molecular weight 515.03 g/mol; formula C29H32F2N2O2·HCl.
  • Store desiccated at room temperature; handle solvents per safety guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.