Learn More
Takara Bio In-Fusion Snap Assembly Value Bundle | 50 rxns
SDP
Catalog No. NC1877406
Encompass
In-Fusion Snap Assembly Master Mix enables high-efficiency, high-fidelity, directional cloning of one or more PCR fragments into any vector. In addition to the cloning kit, this package includes a NucleoSpin Gell and PCR Clean-Up Kit, Stellar Competent Cells, and PrimeSTAR Max DNA Polymerase.
Catalog No. NC1877406 Supplier Takara Bio Supplier No. 638946
In-Fusion Snap Assembly Master Mix enables high-efficiency, high-fidelity, directional cloning of one or more PCR fragments into any vector. In addition to the cloning kit, this package includes a NucleoSpin Gell and PCR Clean-Up Kit, Stellar Competent Cells, and PrimeSTAR Max DNA Polymerase.