Learn More
New England Biolabs, Inc. SNAP-Biotin – 50 nmol
SDP
Catalog No. 508112053
Encompass
SNAP-Biotin is suitable for applications such as biotinylation of SNAP-tag fusion proteins in living cells for detection with streptavidin fluorophore conjugates or labeling in solution for analysis by SDS-PAGE/Western blot or for capture with streptavidin for binding and interaction studies.
Catalog No. 50-811-2053 Supplier New England Biolabs, Inc. Supplier No. S9110S
SNAP-Biotin is suitable for applications such as biotinylation of SNAP-tag fusion proteins in living cells for detection with streptavidin fluorophore conjugates or labeling in solution for analysis by SDS-PAGE/Western blot or for capture with streptavidin for binding and interaction studies.