Learn More
R&D Systems SNAP BURST CAP 13MM (10/PACK)
SDP
Catalog No. NC9277726
Encompass
Snap burst cap - 13mm with septum/Si, 10/pkg. Suitable for use with the iCE280-PrinCE microinjector configuration only.
Catalog No. NC9277726 Supplier R&D Systems Supplier No. 102032/DEL
Customers require a valid business account associated to their profile to access pricing and ordering options.
Snap burst cap - 13mm with septum/Si, 10/pkg. Suitable for use with the iCE280-PrinCE microinjector configuration only.