Learn More
Electron Microscopy Sciences MacroTubes™ with Snap-Caps, Blue
SDP
Catalog No. 501929147
Encompass_Preferred
The 5 ml MacroTubes thave all conveniences of standard 1.5 ml tubes but are scaled up for larger volumes. The attached cap provides a tight, leak proof seal from -86 deg C to +80 deg C and can be opened or closed with one hand. The tubes are made of maximum clarity, low-binding virgin polypropylene and are certified RNase and DNase free.
Tubes are sized to be compatible with all Benchmark (USA) and standard German-designed equipment. The 16 mm tube diameter is identical to the diameter of 15 ml tubes, ensuring compatibility.
The potential contamination of pipettor shafts is greatly reduced because the 56 mm total tube length (vs. 120 mm for 15 ml tubes) is short enough for standard 1 ml or 5 ml tips to reach the tube's conical bottom. Feature dual graduations at every 0.2 ml and 0.5 ml.
Color: Blue
Catalog No. 50-192-9147 Supplier Electron Microscopy Sciences Supplier No. 6302503
The 5 ml MacroTubes thave all conveniences of standard 1.5 ml tubes but are scaled up for larger volumes. The attached cap provides a tight, leak proof seal from -86 deg C to +80 deg C and can be opened or closed with one hand. The tubes are made of maximum clarity, low-binding virgin polypropylene and are certified RNase and DNase free.
Tubes are sized to be compatible with all Benchmark (USA) and standard German-designed equipment. The 16 mm tube diameter is identical to the diameter of 15 ml tubes, ensuring compatibility.
The potential contamination of pipettor shafts is greatly reduced because the 56 mm total tube length (vs. 120 mm for 15 ml tubes) is short enough for standard 1 ml or 5 ml tips to reach the tube's conical bottom. Feature dual graduations at every 0.2 ml and 0.5 ml.
Color: Blue