Learn More
Electron Microscopy Sciences Five-O™ 5ml Tubes Attchd Screw Caps, Sterile
SDP
Catalog No. 501929154
Encompass_Preferred
Our new 5 ml Five-O screw cap tubes are made of maximim clarity, low-binding virgin polypropylene and are certified RNase and DNase free. They are sized to be compatible with all Benchmark (USA) and standard German-designed equipment, including centrifuges, dry bath block heaters, tube racks and thermal shakers/vortexers. Screw cap tubes are supplied with 20 mm OD caps that seal smoothly and completely with a single 360 degree turn.
Useful from -86 deg C to 100 deg C, they are a valuable resource for boiling and sample preservation at very low temperatures. The foam racks are similar to the packaging normally associated with 15 ml tubes (upright woth caps attached), but require about 60 percent less storage space. Feature dual graduations at every 0.2 ml and 0.5 ml.
Sterile: Sterile
Catalog No. 50-192-9154 Supplier Electron Microscopy Sciences Supplier No. 6302510
Our new 5 ml Five-O screw cap tubes are made of maximim clarity, low-binding virgin polypropylene and are certified RNase and DNase free. They are sized to be compatible with all Benchmark (USA) and standard German-designed equipment, including centrifuges, dry bath block heaters, tube racks and thermal shakers/vortexers. Screw cap tubes are supplied with 20 mm OD caps that seal smoothly and completely with a single 360 degree turn.
Useful from -86 deg C to 100 deg C, they are a valuable resource for boiling and sample preservation at very low temperatures. The foam racks are similar to the packaging normally associated with 15 ml tubes (upright woth caps attached), but require about 60 percent less storage space. Feature dual graduations at every 0.2 ml and 0.5 ml.
Sterile: Sterile