Learn More
Krackeler Scientific Snap Caps with 11mm Septa
SDP
Catalog No. 51820550N
Change view
Click to view available options
Color:
Clear
Material (Liner):
PTFE/Silicone Rubber
Catalog No. Color Material (Liner)
51820550N Clear PTFE/Silicone Rubber
1 options
Catalog No. 51820550N Supplier Krackeler Scientific Supplier No. 9251820550EA
Add to Cart
Add to Cart
For vials with wide opening inserts
  • Snap-on closure can be removed and replaced during pipetting.
  • Nonbonded components can be separated for disposal.
Specifications
Material (Liner) PTFE/Silicone Rubber
Color Clear
Material Polyethylene
Pre-slit No
Septum PTFE/Red Silicone
Quantity 100 Pk.
Closure Size 11 mm