Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RayDyLyo™ Snap Caps from DWK Life Sciences™
SDP

Catalog No. 01000891 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
20 mm
Color:
Blue
Packaging Type:
Nested
For Use With (Equipment):
Serum Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size Color Packaging Type For Use With (Equipment)
01-000-891 20 mm Blue Nested Serum Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-000-891 Supplier RayDyLyo™ Supplier No. 255843000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

These plastic push-fit caps are designed for closing vials of injectable drugs. Flip-off snap caps are pre-assembled with an ISO 8362-2 compliant elastomer stopper to simplify the production process.

  • Container Closure Integrity (CCI) validated for use with ISO 8362-1 glass and COP vials
  • Delivered sterile (RTU) with a 2 year shelf life from time of manufacture
  • Clean room manufactured with 100% in-process quality controls
  • Suitable for aseptic small batch, clinical trial, and robotic fill and finish operations
  • Snap caps do not require special crimping tools and affix with single vertical pressure
  • Provides tamper-evident closing and eliminates crimping defects
  • Compatible with gamma and steam terminal sterilization
  • Available in nested, standard bag, and RTP-type bag configurations
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Diameter (Metric) Outer 20 mm
Material Polypropylene
Sterility Sterile
Packaging Type Nested
Quantity 1728/Pk.
Closure Size 20 mm
For Use With (Equipment) Serum Vials
Autoclavable Autoclavable
Certifications/Compliance ISO 8362-2

RayDyLyo™ is a registered trademark of ARaymond.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.