Learn More
New England Biolabs, Inc. SNAP-Capture Magnetic Beads – 2 ml
SDP
Catalog No. 508112070
Encompass
The SNAP-Capture Magnetic Beads are used to selectively immobilize and magnetically separate a SNAP-tag fusion protein from solution using magnetic agarose beads. They are prepared by the coupling of SNAP-tag substrate benzylguanine with highly stable 20-100 um superparamagnetic particles.
Catalog No. 50-811-2070 Supplier New England Biolabs, Inc. Supplier No. S9145S
The SNAP-Capture Magnetic Beads are used to selectively immobilize and magnetically separate a SNAP-tag fusion protein from solution using magnetic agarose beads. They are prepared by the coupling of SNAP-tag substrate benzylguanine with highly stable 20-100 um superparamagnetic particles.