Learn More
New England Biolabs, Inc. SNAP-Cell 505-Star – 50 nmol
SDP
Catalog No. 508112047
Encompass
SNAP-Cell 505-Star is a photostable green fluorescent substrate that can be used to label SNAP-tag fusion proteins inside living cells, on cell surfaces, or in vitro.
Catalog No. 50-811-2047 Supplier New England Biolabs, Inc. Supplier No. S9103S
SNAP-Cell 505-Star is a photostable green fluorescent substrate that can be used to label SNAP-tag fusion proteins inside living cells, on cell surfaces, or in vitro.