Learn More
New England Biolabs, Inc. SNAP-Cell 647-SiR – 30 nmol
SDP
Catalog No. 501047526
Encompass
SNAP-Cell 647-SiR is a far-red fluorescent substrate that can be used to label SNAP-tag fusion proteins inside living cells, on cell surfaces, or in vitro.
Catalog No. 50-104-7526 Supplier New England Biolabs, Inc. Supplier No. S9102S
SNAP-Cell 647-SiR is a far-red fluorescent substrate that can be used to label SNAP-tag fusion proteins inside living cells, on cell surfaces, or in vitro.