Learn More
New England Biolabs, Inc. SNAP-Cell Block – 100 nmol
SDP
Catalog No. 508112050
Encompass
SNAP-Cell Block (bromothenylpteridine, BTP) is a non-fluorescent compound that blocks the reactivity of the SNAP-tag in solution or in living cells.
Catalog No. 50-811-2050 Supplier New England Biolabs, Inc. Supplier No. S9106S
SNAP-Cell Block (bromothenylpteridine, BTP) is a non-fluorescent compound that blocks the reactivity of the SNAP-tag in solution or in living cells.