Learn More
New England Biolabs, Inc. SNAP-Cell Oregon Green – 50 nmol
SDP
Catalog No. NC1239707
Encompass
SNAP-Cell Oregon Green is a photostable green fluorescent substrate that can be used to label SNAP-tag fusion proteins inside living cells or in vitro. Conjugates of Oregon Green are more photostable than those of fluorescein, and their fluorescence properties are essentially pH insensitive in the physiological pH range.
Catalog No. NC1239707 Supplier New England Biolabs, Inc. Supplier No. S9104S
SNAP-Cell Oregon Green is a photostable green fluorescent substrate that can be used to label SNAP-tag fusion proteins inside living cells or in vitro. Conjugates of Oregon Green are more photostable than those of fluorescein, and their fluorescence properties are essentially pH insensitive in the physiological pH range.