Learn More
Neta Scientific Agilent Snap cap with red silicone septa 100/PK
SDP
Supplier:  Neta Scientific 51820550
Encompass_Preferred
This item is manufactured by Agilent
Agilent Cap, snap, clear, clear PTFE/red silicone septa, 100/pk. Cap size: 11 mm - 2 mL Crimp/Snap Top Vials & Caps
UNSPSC Code  41121800
- 58
Catalog No. 50-929-166