Learn More
MSC Rochester Midland Corporation All-Purpose Cleaner, Disinfectant, Liquid, Floral, 0.5 gal
Catalog No. 502816592
Encompass
This all-purpose cleaner is a liquid disinfectant with a floral scent, designed for use on hard surfaces. It is GreenGuard Certified, Leadership in Energy & Environmental Design Certified, and Biodegradable.
  • Hard water tolerance
  • NPE Free
  • Organic soil tolerance
  • Neutral formula will not dull floor finishes
  • Cleans, disinfects, deodorizes
  • Pseudomonicidal, fungicidal, virucidal
Catalog No. 50-281-6592 Supplier MSC Supplier No. 01355825
This all-purpose cleaner is a liquid disinfectant with a floral scent, designed for use on hard surfaces. It is GreenGuard Certified, Leadership in Energy & Environmental Design Certified, and Biodegradable.
  • Hard water tolerance
  • NPE Free
  • Organic soil tolerance
  • Neutral formula will not dull floor finishes
  • Cleans, disinfects, deodorizes
  • Pseudomonicidal, fungicidal, virucidal