Learn More
IDEXX REFERENCE LABORATORIES SNAP FIV/FELV COMBO TEST 30 PK
Supplier:  IDEXX REFERENCE LABORATORIES 9908354
Encompass
This supplier will only sell to Pharmaceutical and Manufacturing companies. They will not ship to Universities or Hospitals.
SNAP FIV/FeLV Combo TestScreen for two infectious diseases with one sample in just 10 minutes.Accurately screen for FIV and FeLV.Prevent the spread of these diseases while increasing awareness of prevalence in your area by offering clients annual testing.Easily implement feline health protocols as recommended by the AAFP (American Association of Feline Practitioners).SNAP tests provide reference laboratory quality technology for superior diagnostic sensitivity and specificity at the point of care.Automatically activate SNAP tests to save time and improve work flow with the SNAP Pro Analyzer.
Catalog No. NC2223230