Learn More
Medchemexpress LLC Snap-fusion protein ligand-1 | 913817-45-5 | 96.0% | 264.71 g/mol | C12H13ClN4O | 250 MG
SDP
Catalog No. 5000283436
Encompass_Preferred
SNAP-fusion protein ligand-1 is a small-molecule ligand designed to bind SNAP-tag fusion proteins and serve as a synthetic handle in PROTAC assembly and other bioconjugation workflows. It is supplied as a solid with batch-specific analytical data and storage recommendations provided by the manufacturer; users should consult the certificate of analysis for exact purity and handling instructions.
  • Used as a ligand for SNAP-tag fusion proteins.
  • Suitable for assembly of proteolysis-targeting chimeras and bioconjugates.
  • Supplied as a solid with specified storage recommendations.
  • Batch-specific purity and analytical data provided in the certificate of analysis.
  • Analytical characterization reported by LCMS.
Catalog No. 50-002-83436 Supplier Medchemexpress LLC Supplier No. HYW128709250MG
SNAP-fusion protein ligand-1 is a small-molecule ligand designed to bind SNAP-tag fusion proteins and serve as a synthetic handle in PROTAC assembly and other bioconjugation workflows. It is supplied as a solid with batch-specific analytical data and storage recommendations provided by the manufacturer; users should consult the certificate of analysis for exact purity and handling instructions.
  • Used as a ligand for SNAP-tag fusion proteins.
  • Suitable for assembly of proteolysis-targeting chimeras and bioconjugates.
  • Supplied as a solid with specified storage recommendations.
  • Batch-specific purity and analytical data provided in the certificate of analysis.
  • Analytical characterization reported by LCMS.