Learn More
Instech Snap-in spring swivel arm for rat shoebox cage top and plastic swivel
SDP
Catalog No. NC1293397
Encompass
The Instech CM375KRP is a snap‑in spring swivel mount designed for use with plastic swivels in rat shoebox cages. Its flexible spring counterbalances the weight of the swivel and tether to prevent slack as the animal moves, improving mobility and reducing strain. Swivels snap into or out of the arm with one hand and no tools, and the mount is compatible with slotted cage lids to allow animal transfer without disconnecting the tether.
Features
  • Snap‑in spring swivel mount for plastic swivels
  • Flexible spring counterbalances swivel/tether weight
  • Prevents slack as animals move
  • One‑handed swivel insertion/removal
  • Use with slotted shoebox cage lids
  • Safe for cage washers
Catalog No. NC1293397 Supplier Instech Supplier No. CM375KRP
The Instech CM375KRP is a snap‑in spring swivel mount designed for use with plastic swivels in rat shoebox cages. Its flexible spring counterbalances the weight of the swivel and tether to prevent slack as the animal moves, improving mobility and reducing strain. Swivels snap into or out of the arm with one hand and no tools, and the mount is compatible with slotted cage lids to allow animal transfer without disconnecting the tether.
Features
  • Snap‑in spring swivel mount for plastic swivels
  • Flexible spring counterbalances swivel/tether weight
  • Prevents slack as animals move
  • One‑handed swivel insertion/removal
  • Use with slotted shoebox cage lids
  • Safe for cage washers