Learn More
StatLab Snap n' Digest Tuberculosis Collection Kit
SDP
Catalog No. 22130485
Change view
Click to view available options
Volume (Metric):
75 mL
150 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-130-485 150 mL
22-130-484 75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-130-485 Supplier StatLab Supplier No. 668
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Ready-to-mix system to process Tuberculosis and other clinical specimens (sputum, bronchial, tracheal lavage) suspected to contain mycobacteria.
  • Contains crushable NALC ampoules inside a safety straw inside a flexible bottle with alkali solution and TB phosphate buffer packets.
  • Compatible with multiple automated TB systems
  • Solution in bottle with unbroken ampoule is stable for 1 year without refrigeration
  • 10 bottles/box
Available in 75 mL or 150 mL:
  • 75 mL contains 1 NALC ampoule, 4 oz. bottle with 2 oz. of alkali solution, and 5 TB Phosphate Buffer packets
  • 150 mL contains 2 NALC ampoules, 8 oz. bottle with 4 oz. of alkali solution, and 10 TB Phosphate TB Buffer packets
Specifications
Product Type Collection and Transport System
Format Tube
Sterility Non-sterile
Quantity 10 Bottles/Box
Volume (Metric) 150 mL