Learn More
StatLab Snap n' Stain Iron Stain Dropper
SDP
Catalog No. 22130483
Change view
Click to view available options
Snap n' Stain Iron Stain Dropper
Catalog No.
22-130-483 Snap n' Stain Iron Stain Dropper
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-130-483 Supplier StatLab Supplier No. 661
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Easy-to-Use Dropper
  • Each dropper unit contains two reagent-filled glass ampoules
  • Just break and mix
  • Can stain up to four slides
  • 50 droppers/box
Specifications
Quantity 50/Pk.
Product Type Iron Stain