Learn More
Abcam SNAP, NO donor, 10MG
Catalog No. 502658338
Encompass_Preferred
MW 220.25 Da. NO donor. Vasodilator in vitro and in vivo, that does not induce pharmacological tolerance.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-8338 Supplier Abcam Supplier No. AB12001410MG
MW 220.25 Da. NO donor. Vasodilator in vitro and in vivo, that does not induce pharmacological tolerance.
The product is subject to the following: Abcam Restricted Use Statement