Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Snap Oa Reagent

Catalog No. 7989165G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-891-65G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 79-891-65G Supplier Sigma Aldrich Supplier No. 7989165G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

SnAP Reagents provide a one-step route, in tandem with various aldehyde substrates, to saturatedN-heterocycles. The synthesis ofN-heterocycles through SnAP Reagents requires mild reaction conditions, and aldehydes bearing aryl, heteroaryl,glyoxyl,aliphatic, and halogenated groups are well tolerated. This product was introduced in collaboration with theBode Research GroupTechnology spotlight: SnAP Reagents Professor product portal: Jeffrey Bode Research GroupSnAP Reagents for the Synthesis of Piperazines and MorpholinesSnAP reagents for the one-step synthesis of medium-ring saturated N-heterocycles from aldehydesSnAP Reagents for a Cross-Coupling Approach to the One-Step Synthesis of Saturated N-Heterocycles

Specifications

CAS 1313211-51-6
Melting Point 176°C to 181°C
Molecular Formula C11H9N3O3
Linear Formula C11H9N3O3
MDL Number MFCD24386394
Quantity 500 mg
Synonym 4-(4-(Prop-2-yn-1-yloxy)phenyl)-1,2,4-triazolidine-3,5-dione
Molecular Weight (g/mol) 231.21 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.