Learn More
Beckman Coulter Microfuge 18 Centrifuge Tubes, 1.5mL, Natural, Bag of 500
Supplier:  Beckman Coulter 357359
Encompass_Preferred
The Beckman Coulter Microfuge 18 Centrifuge is a compact benchtop centrifuge designed for quick spin downs and separations. It has a maximum speed of 13,300 rpm and maximum RCF of 16,060 x g. The rotor accommodates 18 x 1.5/2.0 mL microcentrifuge tubes. It has a compact footprint, digital display, and 6 acceleration/deceleration rates. This is an ideal personal centrifuge for routine applications requiring high performance in a small package.
Catalog No. NC2221992